4-Methyl-2-(methylthio)pyrimidine ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Methyl-2(methylsulphanyl)pyrimidine

045164-1G 045164-5G 045164-25G
101869-460EA 38.46 USD
101869-460 101869-462 101869-464
4-Methyl-2-(methylthio)pyrimidine ≥97%
4-Methyl-2-(methylthio)pyrimidine

Hazard: Irritant

Formula: C₆H₈N₂S
MW: 140.21 g/mol
Boiling Pt: 78…80 °C (1 mmHg)
MDL Number: MFCD00023242
CAS Number: 14001-63-9

Order Now

Specification Test Results

Boiling point 78-80°C/1mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR