Triethyl orthochloroacetate ≥93%

Supplier: Matrix Scientific

Synonyms: Orthochloroacetic Acid Triethyl Ester, 2-Chloro-1,1,1-triethoxyethane

045143-1G 045143-5G
101869-342EA 73.85 USD
101869-342 101869-344
Triethyl orthochloroacetate ≥93%
Triethyl orthochloroacetate

Hazard: Irritant

Formula: C₈H₁₇ClO₃
MW: 196.67 g/mol
Boiling Pt: 88…90 °C (20 mmHg)
MDL Number: MFCD00142889
CAS Number: 51076-95-0

Order Now

Specification Test Results

Boiling point 88-90°C/20m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR