1,1,1-Trifluoro-4-methoxy-3-penten-2-one ≥95%

Supplier: Matrix Scientific

045010-500MG 045010-1G 045010-5G
101868-704EA 100 USD
101868-704 101868-706 101868-708
1,1,1-Trifluoro-4-methoxy-3-penten-2-one ≥95%
1,1,1-Trifluoro-4-methoxy-3-penten-2-one

Hazard: Irritant

Formula: C₆H₇F₃O₂
MW: 168.12 g/mol
Boiling Pt: 41°C/9mm
MDL Number: MFCD00173110
CAS Number: 102145-82-4

Order Now

Specification Test Results

Boiling point 41°C/9mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR