3-Chloro-4-phenyl-1,2,5-thiadiazole ≥95%

Supplier: Matrix Scientific

044797-500MG 044797-1G 044797-5G
101867-644EA 375.38 USD
101867-644 101867-646 101867-648
3-Chloro-4-phenyl-1,2,5-thiadiazole ≥95%
3-Chloro-4-phenyl-1,2,5-thiadiazole

Hazard: Irritant

Formula: C₈H₅ClN₂S
Boiling Pt: 94-96°C/0.1mm
MDL Number: MFCD01935960
CAS Number: 5728-14-3

Order Now

Specification Test Results

Boiling point 94-96°C/0.1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR