(6-Chloro-3-pyridinyl)methyl acetate ≥95%

Supplier: Matrix Scientific

044646-500MG 044646-1G 044646-5G
101866-888EA 278.46 USD
101866-888 101866-890 101866-892
(6-Chloro-3-pyridinyl)methyl acetate ≥95%
(6-Chloro-3-pyridinyl)methyl acetate

Hazard: Irritant

Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Boiling Pt: 92 °C (0.01 mmHg)
MDL Number: MFCD04973731
CAS Number: 881734-66-3

Order Now

Specification Test Results

Boiling point 92°C/0.01mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR