2-Chloro-6-methyl-4-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

002791-1G 002791-5G
101784-668EA 44.62 USD
101784-668 101784-670
2-Chloro-6-methyl-4-(trifluoromethyl)pyridine ≥97%
2-Chloro-6-methyl-4-(trifluoromethyl)pyridine

Hazard: Flammable

Formula: C₇H₅ClF₃N
Boiling Pt: 51-52°C/15mm
MDL Number: MFCD00052900
CAS Number: 22123-14-4

Order Now

Specification Test Results

Boiling point 51-52°C/15m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR