2-Chloro-6-fluorophenylacetonitrile

Supplier: Matrix Scientific

002780-5G 002780-25G
101784-638EA 86.15 USD
101784-638 101784-640
2-Chloro-6-fluorophenylacetonitrile
2-Chloro-6-fluorophenylacetonitrile

Hazard: Toxic

Formula: C₈H₅ClFN
MW: 169.59 g/mol
Boiling Pt: 85°C/2mm
Melting Pt: 40-43°C
MDL Number: MFCD00001900
CAS Number: 75279-55-9
UN: 3439
ADR: 6.1,II

Order Now

Specification Test Results

Boiling point 85°C/2mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR