Methyl-3,5-bis(trifluoromethyl)benzoate ≥98%

Supplier: Matrix Scientific

Synonyms: Methyl 3,5-bis(trifluoromethyl)benzoate

002681-5G 002681-25G 002681-100G
101784-422EA 13.85 USD
101784-422 101784-424 101784-426
Methyl-3,5-bis(trifluoromethyl)benzoate ≥98%
Methyl 3,5-bis(trifluoromethyl)benzoate

Hazard: Irritant

Formula: C₁₀H₆F₆O₂
Boiling Pt: 85…87 °C (15 mmHg)
MDL Number: MFCD00052317
CAS Number: 26107-80-2

Order Now

Specification Test Results

Boiling point 85-87°C/15m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR