α-Methyl-4-trifluoromethylbenzyl alcohol

Supplier: Matrix Scientific

Synonyms: 1-[4-(Trifluoromethyl)phenyl]ethanol

002606-1G 002606-5G
101784-276EA 20 USD
101784-276 101784-278
α-Methyl-4-trifluoromethylbenzyl alcohol
alpha-Methyl-4-trifluoromethylbenzyl alcohol

Hazard: Irritant

Formula: C₉H₉F₃O
Boiling Pt: 143-145°C/20mm
MDL Number: MFCD00039618
CAS Number: 1737-26-4

Order Now

Specification Test Results

Boiling point 143-145°C/2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR