2-Chloro-4-methylanisole ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-1-methoxy-4-methylbenzene, 3-Chloro-4-methoxytoluene

002563-5G 002563-25G
101784-172EA 116.92 USD
101784-172 101784-174
2-Chloro-4-methylanisole ≥98%
2-Chloro-4-methylanisole

Hazard: Irritant

Formula: C₈H₉ClO
MW: 156.609 g/mol
Boiling Pt: 100°C/22mm
MDL Number: MFCD00060649
CAS Number: 22002-44-4

Order Now

Specification Test Results

Boiling point 100°C/22mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR