α-Methyl-3-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: Matrix Scientific

Synonyms: 1-[3-(Trifluoromethyl)phenyl]ethanol

002399-5G 002399-25G
101783-838EA 72.31 USD
101783-838 101783-840
α-Methyl-3-(trifluoromethyl)benzyl alcohol ≥97%
alpha-Methyl-3-(trifluoromethyl)benzyl alcohol

Hazard: Irritant

Formula: C₉H₉F₃O
Boiling Pt: 89-91°C/4mm
Flash Pt: 96°C
MDL Number: MFCD00021868
CAS Number: 454-91-1

Order Now

Specification Test Results

Boiling point 89-91°C/4mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR