5-Fluoro-3-methylbenzo[b]thiophene

Supplier: Matrix Scientific

002383-1G 002383-5G
101783-808EA 44.62 USD
101783-808 101783-810
5-Fluoro-3-methylbenzo[b]thiophene
5-Fluoro-3-methylbenzo[b]thiophene

Hazard: Irritant

Formula: C₉H₇FS
Boiling Pt: 68°C/0.5mm
MDL Number: MFCD00179348
CAS Number: 17514-63-5

Order Now

Specification Test Results

Boiling point 68°C/0.5mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR