5-Chloro-2-methylbenzylamine

Supplier: Matrix Scientific

002173-1G 002173-5G
101783-386EA 36.92 USD
101783-386 101783-388
5-Chloro-2-methylbenzylamine
5-Chloro-2-methylbenzylamine

Hazard: Irritant

Formula: C₈H₁₀ClN
MW: 155.625 g/mol
Boiling Pt: 100°C/10mm
MDL Number: MFCD01310800
CAS Number: 27917-13-1

Order Now

Specification Test Results

Boiling point 100°C/10mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR