3-(1,1,2,2-Tetrafluoroethoxy)nitrobenzene ≥98%

Supplier: Matrix Scientific

002039-5G 002039-25G
101783-088EA 63.08 USD
101783-088 101783-090
3-(1,1,2,2-Tetrafluoroethoxy)nitrobenzene ≥98%
3-(1,1,2,2-Tetrafluoroethoxy)nitrobenzene

Hazard: Irritant

Formula: C₈H₅F₄NO₃
Boiling Pt: 72…74 °C (0.1 mmHg)
CAS Number: 1644-21-9

Order Now

Specification Test Results

Boiling point 72-74°C/0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR