Allyl 1H,1H-heptafluorobutyl ether ≥97%
Supplier: Matrix Scientific
001927-5G
001927-25G
101782-862EA
76.92
USD
101782-862
101782-864
Allyl 1H,1H-heptafluorobutyl ether ≥97%
Allyl 1H,1H-heptafluorobutyl ether
Hazard: Stored under Argon
Formula:
C₇H₇F₇O Boiling Pt: 105 °C Density: 1.31 |
MDL Number:
MFCD01862010 CAS Number: 648-42-0 |
Specification Test Results
Boiling point | 105°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...