Methyl-2,5-bis(trifluoromethyl)-3,6-dioxaundecafluorononanoate (mixture of isomers) ≥97%

Supplier: Matrix Scientific

Synonyms: 2,5-Bis(trifluoromethyl)-3,6-dioxaundecafluorononanoic Acid Methyl Ester

001420-5G 001420-25G
101781-620EA 78.46 USD
101781-620 101781-622
Methyl-2,5-bis(trifluoromethyl)-3,6-dioxaundecafluorononanoate (mixture of isomers) ≥97%
Methyl-2,5-bis(trifluoromethyl)-3,6-dioxaundecafluorononanoate (mixture of isomers)

Hazard: Irritant

Formula: C₁₀H₃F₁₇O₄
MW: 510.10 g/mol
Boiling Pt: 155-156°C
MDL Number: MFCD00054723
CAS Number: 26131-32-8

Order Now

Specification Test Results

Boiling point 155-156°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR