1-Bromo-2-(bromodifluoromethyl)cyclohexane ≥97%

Supplier: Matrix Scientific

001324-1G 001324-5G
101781-402EA 29.23 USD
101781-402 101781-404
1-Bromo-2-(bromodifluoromethyl)cyclohexane ≥97%
1-Bromo-2-(bromodifluoromethyl)cyclohexane

Hazard: Irritant

Formula: C₇H₁₀Br₂F₂
Boiling Pt: 108…110 °C (40 mmHg)
MDL Number: MFCD00077459
CAS Number: 14737-09-8

Order Now

Specification Test Results

Boiling point 108-110°C/4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR