4,4,5,5,6,6,6-Heptafluoro-1-phenyl-1,3-hexanedione ≥97%

Supplier: Matrix Scientific

001298-5G 001298-25G
101781-354EA 141.54 USD
101781-354 101781-356
4,4,5,5,6,6,6-Heptafluoro-1-phenyl-1,3-hexanedione ≥97%
4,4,5,5,6,6,6-Heptafluoro-1-phenyl-1,3-hexanedione

Hazard: Irritant

Formula: C₁₂H₇F₇O₂
Boiling Pt: 70 °C (0.1 mmHg)
MDL Number: MFCD00511283
CAS Number: 53580-21-5

Order Now

Specification Test Results

Boiling point 70°C/0.1mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR