1,1,1,3,5,5,5-Heptafluoropentan-2,4-dione ≥97%

Supplier: Matrix Scientific

001258-1G 001258-5G
101781-256EA 136.92 USD
101781-256 101781-258
1,1,1,3,5,5,5-Heptafluoropentan-2,4-dione ≥97%
1,1,1,3,5,5,5-Heptafluoropentan-2,4-dione

Hazard: Corrosive

Formula: C₅HF₇O₂
MW: 226.05 g/mol
Boiling Pt: 70…72 °C
MDL Number: MFCD00153187
CAS Number: 77968-17-3

Order Now

Specification Test Results

Boiling point 70-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR