4-Bromo-3,3,4,4-tetrafluoro-1-butanol ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-3,3,4,4-tetrafluorobutan-1-ol

001172-1G 001172-5G
101781-074EA 63.08 USD
101781-074 101781-076
4-Bromo-3,3,4,4-tetrafluoro-1-butanol ≥97%
4-Bromo-3,3,4,4-tetrafluoro-1-butanol

Hazard: Irritant/Light Sensitive

Formula: C₄H₅BrF₄O
MW: 224.99 g/mol
Boiling Pt: 148 °C
MDL Number: MFCD00077467
CAS Number: 234443-21-1

Order Now

Specification Test Results

Boiling point 148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR