1-Bromo-3,3,4,4,4-pentafluoro-2-butanone ≥97%
Supplier: Matrix Scientific
001170-1G
001170-5G
101781-070EA
56.92
USD
101781-070
101781-072
1-Bromo-3,3,4,4,4-pentafluoro-2-butanone ≥97%
1-Bromo-3,3,4,4,4-pentafluoro-2-butanone
Hazard: Corrosive/Lachrymator
Formula:
C₄H₂BrF₅O MW: 240.96 Boiling Pt: 93…94 °C |
MDL Number:
MFCD00041447 CAS Number: 92737-01-4 UN: 3265 ADR: 8,II |
Specification Test Results
Boiling point | 93-94°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...