(1H,1H,2H,2H-Perfluoro-n-hexyl)methyldichlorosilane ≥97%

Supplier: Matrix Scientific

001157-5G 001157-25G
101781-044EA 207.69 USD
101781-044 101781-046
(1H,1H,2H,2H-Perfluoro-n-hexyl)methyldichlorosilane ≥97%
(1H,1H,2H,2H-Perfluoro-n-hexyl)methyldichlorosilane

Hazard: Flammable/Corrosive

Formula: C₇H₇Cl₂F₉Si
Boiling Pt: 86°C/48mm
MDL Number: MFCD00077577
CAS Number: 38436-16-7

Order Now

Specification Test Results

Boiling point 86°C/48mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR