1-Bromo-1,1-difluoro-2-heptene (E/Z) (E/Z=95:5) ≥97%

Supplier: Matrix Scientific

001135-1G 001135-5G
101781-026EA 253.85 USD
101781-026 101781-028
1-Bromo-1,1-difluoro-2-heptene (E/Z) (E/Z=95:5) ≥97%
1-Bromo-1,1-difluoro-2-heptene (E/Z)

Hazard: Irritant

Formula: C₇H₁₁BrF₂
Boiling Pt: 78-87°C/60mm
MDL Number: MFCD00153694
CAS Number: 262296-38-8

Order Now

Specification Test Results

Boiling point 78-87°C/60m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR