2-Bromo-1,1,2-trifluoroethyl ethyl ether ≥97%

Supplier: Matrix Scientific

001131-5G 001131-25G
101781-016EA 130.77 USD
101781-016 101781-018
2-Bromo-1,1,2-trifluoroethyl ethyl ether ≥97%
2-Bromo-1,1,2-trifluoroethyl ethyl ether

Hazard: Irritant/Refrigerate

Formula: C₄H₆BrF₃O
Boiling Pt: 116°C
MDL Number: MFCD00042465
CAS Number: 380-78-9

Order Now

Specification Test Results

Boiling point 116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR