Methyl-3-trifluoromethylbenzoylacetate

Supplier: Matrix Scientific

Synonyms: Methyl 3-oxo-3-[3-(trifluoromethyl)phenyl]propanoate

001108-1G 001108-5G
101780-978EA 41.54 USD
101780-978 101780-980
Methyl-3-trifluoromethylbenzoylacetate
Methyl-3-trifluoromethylbenzoylacetate

Hazard: Irritant

Formula: C₁₁H₉F₃O₃
MW: 246.185 g/mol
Boiling Pt: 94…96 °C (0.2 mmHg)
MDL Number: MFCD00216522
CAS Number: 93618-66-7

Order Now

Specification Test Results

Boiling point 94-96°C/0.2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR