Perfluoro(4-methylpent-2-ene) ≥90%

Supplier: Matrix Scientific

001084-25G 001084-100G 001084-500G
101780-912EA 26.15 USD
101780-912 101780-914 101780-916
Perfluoro(4-methylpent-2-ene) ≥90%
Perfluoro(4-methylpent-2-ene)

Hazard: Irritant/Refrigerate

Formula: C₆F₁₂
Boiling Pt: 49 °C
MDL Number: MFCD00153253
CAS Number: 2070-70-4

Order Now

Specification Test Results

Boiling point 49°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR