2-Iodo-3-methylpyridine ≥97%

Supplier: Matrix Scientific

032532-1G 032532-5G
101833-934EA 36.92 USD
101833-934 101833-936
2-Iodo-3-methylpyridine ≥97%
2-Iodo-3-methylpyridine

Hazard: Irritant/Refrigerate

Formula: C₆H₆IN
Boiling Pt: 80 °C (0.3 mmHg)
MDL Number: MFCD04039350
CAS Number: 22282-58-2

Order Now

Specification Test Results

Boiling point 80°C/0.3mm
ND20 1.625


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR