1-Boc-piperidine ≥99%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl Piperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)piperidine, Piperidine-1-carboxylic acid tert-Butyl ester, tert-butyl piperidine-1-carboxylate, 1-Boc-piperidine
020024-5G
020024-25G
101809-258EA
27.69
USD
101809-258
101809-260
1-Boc-piperidine ≥99%
1-Boc-piperidine
Hazard: Irritant/Store in Freezer
Formula:
C₁₀H₁₉NO₂ MW: 185.27 g/mol Boiling Pt: 130…140 °C (15 mmHg) Density: 0.965 Flash Pt: 86 °C |
MDL Number:
MFCD02093935 CAS Number: 75844-69-8 |
Specification Test Results
Boiling point | 130-140°C/1 |
ND20 | 1.4535 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...