2,6-Dichloro-3,5-difluorophenol

Supplier: Matrix Scientific

007423-5G 007423-25G
101792-720EA 29.23 USD
101792-720 101792-722
2,6-Dichloro-3,5-difluorophenol
2,6-Dichloro-3,5-difluorophenol

Hazard: Irritant

Formula: C₆H₂Cl₂F₂O
Boiling Pt: 87…88 °C
MDL Number: MFCD03424471
CAS Number: 63418-08-6

Order Now

Specification Test Results

Boiling point 87-88°C
ND20 1.531

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR