3-Amino-4-(methylthio)benzotrifluoride ≥97%

Supplier: Matrix Scientific

007055-1G 007055-5G
101791-914EA 33.85 USD
101791-914 101791-916
3-Amino-4-(methylthio)benzotrifluoride ≥97%
3-Amino-4-(methylthio)benzotrifluoride

Hazard: Irritant/Irritant-Harmful

Formula: C₈H₈F₃NS
Boiling Pt: 111…112 °C (12 mmHg)
MDL Number: MFCD00052715
CAS Number: 207974-07-0
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 111-112°C/1
ND20 1.537

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR