6-Bromo-1,4-benzodioxane ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-2,3-dihydro-1,4-benzodioxine, 6-Bromo-2,3-dihydrobenzo[b][1,4]dioxine

006761-1G 006761-5G
101791-036EA 38.46 USD
101791-036 101791-038
6-Bromo-1,4-benzodioxane ≥97%
6-Bromo-1,4-benzodioxane

Hazard: Irritant

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 62-64°C/0.2mm
MDL Number: MFCD00040750
CAS Number: 52287-51-1

Order Now

Specification Test Results

Boiling point 62-64°C/0.2
ND20 1.589

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR