β-Trifluoromethylcrotonic acid ≥97%

Supplier: Matrix Scientific

006030-5G 006030-25G
101789-952EA 186.15 USD
101789-952 101789-954
β-Trifluoromethylcrotonic acid ≥97%
beta-Trifluoromethylcrotonic acid

Hazard: Corrosive/Irritant

Formula: C₅H₅F₃O₂
MW: 154.09 g/mol
Boiling Pt: 158-163°C
Melting Pt: 28…30 °C
Flash Pt: 79°C
MDL Number: MFCD01457157
CAS Number: 93404-33-2

Order Now

Specification Test Results

Boiling point 158-163°C
ND20 1.3838

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR