2,3-Dibromohexafluoro-2-butene ≥97%
Supplier: Matrix Scientific
004579-1G
004579-5G
101787-520EA
53.85
USD
101787-520
101787-522
2,3-Dibromohexafluoro-2-butene ≥97%
2,3-Dibromohexafluoro-2-butene
Hazard: Irritant
Formula:
C₄Br₂F₆ MW: 321.85 g/mol Boiling Pt: 104…106 °C |
MDL Number:
MFCD01862090 CAS Number: 384-51-0 |
Specification Test Results
Boiling point | 104-106°C |
ND20 | 1.416 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...