Bis(2-bromoethyl)ether ≥96% stabilised
Supplier: Matrix Scientific
Synonyms:
1-Bromo-2-(2-bromoethoxy)ethane, 2-Bromoethyl ether
Stabilization:
moistured with water
004576-25G
004576-100G
101787-510EA
212.31
USD
101787-510
101787-512
Bis(2-bromoethyl)ether ≥96% stabilised
Bis(2-bromoethyl)ether
TCA listed: T
Hazard: Irritant
Formula:
C₄H₈Br₂O Boiling Pt: 92-93°C/12mm Density: 1.845 Flash Pt: 85°C |
MDL Number:
MFCD00039196 CAS Number: 5414-19-7 UN: 2810 ADR: 6.1,II |
Specification Test Results
Boiling point | 92-93°C/12m |
ND20 | 1.513 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...