1,1-Dibromo-3,3,4,4,4-pentafluoro-2-butanone ≥85%
Supplier: Matrix Scientific
004544-1G
004544-5G
101787-444EA
27.69
USD
101787-444
101787-446
1,1-Dibromo-3,3,4,4,4-pentafluoro-2-butanone ≥85%
1,1-Dibromo-3,3,4,4,4-pentafluoro-2-butanone
Hazard: Corrosive/Lachrymator
Formula:
C₄HBr₂F₅O Boiling Pt: 123°C |
CAS Number:
203302-96-9 |
Specification Test Results
Boiling point | 123°C |
ND20 | 1.3983 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...