Ethyl-3-amino-4,4,4-trifluorocrotonate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-amino-4,4,4-trifluorobut-2-enoate

004515-25G 004515-100G
101787-368EA 49.23 USD
101787-368 101787-370
Ethyl-3-amino-4,4,4-trifluorocrotonate ≥97%
Ethyl 3-amino-4,4,4-trifluorocrotonate

Hazard: Corrosive/Store in Freezer

Formula: C₆H₈F₃NO₂
MW: 183.13 g/mol
Boiling Pt: 83-85°C/15mm
Melting Pt: 26°C
Density: 1.245
MDL Number: MFCD00068195
CAS Number: 372-29-2

Order Now

Specification Test Results

Boiling point 83-85°C/15m
ND20 1.4213

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR