4,4,5,5,6,6,6-Heptafluorohexane-1,2-diol ≥97%

Supplier: Matrix Scientific

008981-1G 008981-5G
101796-008EA 60 USD
101796-008 101796-010
4,4,5,5,6,6,6-Heptafluorohexane-1,2-diol ≥97%
4,4,5,5,6,6,6-Heptafluorohexane-1,2-diol

Hazard: Irritant

Formula: C₆H₇F₇O₂
Boiling Pt: 114 °C (22 mmHg)
MDL Number: MFCD03424477
CAS Number: 1992-91-2

Order Now

Specification Test Results

Boiling point 114°C/22mm
ND20 1.3516


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR