n-Butyl 2,2,2-trifluoroethylacetaldehyde acetal ≥95%

Supplier: Matrix Scientific

Synonyms: Acetaldehyde butyl 2,2,2-trifluoroethyl acetal

008865-5G 008865-25G
101795-660EA 63.08 USD
101795-660 101795-662
n-Butyl 2,2,2-trifluoroethylacetaldehyde acetal ≥95%
n-Butyl 2,2,2-trifluoroethylacetaldehyde acetal

Hazard: Irritant/Irritant-Harmful

Formula: C₈H₁₅F₃O₂
MW: 200.2 g/mol
Boiling Pt: 143-144°C/751mm
MDL Number: MFCD03094109
CAS Number: 2925-42-0

Order Now

Specification Test Results

Boiling point 143-144°C/7
ND20 1.3624

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR