1H,1H,2H,2H-Perfluorooctyl isobutyrate ≥97%
Supplier: Matrix Scientific
002241-1G
002241-5G
101783-512EA
53.85
USD
101783-512
101783-514
1H,1H,2H,2H-Perfluorooctyl isobutyrate ≥97%
1H,1H,2H,2H-Perfluorooctyl isobutyrate
Hazard: Irritant
Formula:
C₁₂H₁₁F₁₃O₂ Boiling Pt: 92 °C (14 mmHg) |
MDL Number:
MFCD01862031 CAS Number: 242812-05-1 |
Specification Test Results
Boiling point | 92°C/14mm |
ND20 | 1.426 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...