Imidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

066643-1G 066643-5G
101932-052EA 23.08 USD
101932-052 101932-054
Imidazo[1,2-a]pyridine ≥95%
Imidazo[1,2-a]pyridine

Hazard: Irritant/Refrigerate

Formula: C₇H₆N₂
MW: 118.1359 g/mol
Boiling Pt: 103°C/1mm
Density: 1.165
MDL Number: MFCD00005553
CAS Number: 274-76-0

Order Now

Specification Test Results

Boiling point 103°C/1mm
Density 1.165

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR