Ethyl-1-naphthoate ≥99%

Supplier: Matrix Scientific

057073-10G 057073-50G
101912-264EA 112.31 USD
101912-264 101912-266
Ethyl-1-naphthoate ≥99%
Ethyl-1-naphthoate

Hazard: Irritant

Formula: C₁₃H₁₂O₂
Boiling Pt: 308-310°C
Density: 1.11
MDL Number: MFCD00016805
CAS Number: 3007-97-4

Order Now

Specification Test Results

Boiling point 308-310°C
Density 1.11


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR