3,4-Xylylacetonitrile

Supplier: Matrix Scientific

057042-10G 057042-50G
101912-148EA 792.31 USD
101912-148 101912-150
3,4-Xylylacetonitrile
3,4-Xylylacetonitrile

Hazard: Irritant

Formula: C₁₀H₁₁N
Boiling Pt: 86°C/1mm
Melting Pt: 35-38°C
Density: 0.99
MDL Number: MFCD00019829
CAS Number: 3020-06-2

Order Now

Specification Test Results

Boiling point 86°C/1mm
Density 0.99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR