2,4-Dimethoxytoluene ≥99%

Supplier: Matrix Scientific

Synonyms: 2,4-Dimethoxy-1-methylbenzene

057031-5G 057031-25G
101912-108EA 453.85 USD
101912-108 101912-110
2,4-Dimethoxytoluene ≥99%
2,4-Dimethoxytoluene

Hazard: Irritant

Formula: C₉H₁₂O₂
MW: 152.1904 g/mol
Boiling Pt: 110-120°C/30mm
Density: 1.04
MDL Number: MFCD00010749
CAS Number: 38064-90-3

Order Now

Specification Test Results

Boiling point 110-120°C/3
Density 1.04

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR