4-Bromo-2,3-dimethylanisole ≥98%
Supplier: Matrix Scientific
Synonyms:
1-Bromo-4-methoxy-2,3-dimethylbenzene
056911-5G
056911-25G
101911-694EA
156.92
USD
101911-694
101911-696
4-Bromo-2,3-dimethylanisole ≥98%
4-Bromo-2,3-dimethylanisole
Hazard: Irritant
Formula:
C₉H₁₁BrO MW: 215.0900 g/mol Boiling Pt: 112°C/5mm Density: 1.38 |
MDL Number:
MFCD01652199 CAS Number: 50638-48-7 |
Specification Test Results
Boiling point | 112°C/5mm |
Density | 1.38 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...