3-Bromobenzyl thiocyanate ≥98%
Supplier: Matrix Scientific
056903-10G
056903-50G
101911-660EA
140
USD
101911-660
101911-662
3-Bromobenzyl thiocyanate ≥98%
3-Bromobenzyl thiocyanate
Hazard: Irritant-Harmful
Formula:
C₈H₆BrNS Boiling Pt: 112 °C (1 mmHg) Density: 1.53 |
MDL Number:
MFCD09025660 CAS Number: 1206228-39-8 |
Specification Test Results
Boiling point | 112°C/1mm |
Density | 1.53 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...