Propargyl isothiocyanate ≥95%

Supplier: Matrix Scientific

056620-1G 056620-5G
101911-066EA 207.69 USD
101911-066 101911-068
Propargyl isothiocyanate ≥95%
Propargyl isothiocyanate

Hazard: Irritant-Harmful/Refrigerate/Lachrymator

Formula: C₄H₃NS
Boiling Pt: 43°C/15 Torr
Density: 1.06
MDL Number: MFCD00060417
CAS Number: 54122-88-2

Order Now

Specification Test Results

Boiling point 43°C/15mm
Density 1.06


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR