2-(1-Piperidino)ethyl isothiocyanate ≥98%

Supplier: Matrix Scientific

056619-1G 056619-5G
101911-062EA 56.92 USD
101911-062 101911-064
2-(1-Piperidino)ethyl isothiocyanate ≥98%
2-(1-Piperidino)ethyl isothiocyanate

Hazard: Toxic/Corrosive/Moisture Sensitive

Formula: C₈H₁₄N₂S
MW: 170.28 g/mol
Boiling Pt: 84°C/5mm
Density: 1.04
MDL Number: MFCD00041232
CAS Number: 32813-24-4

Order Now

Specification Test Results

Boiling point 84°C/5mm
Density 1.04

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR