3-Phenylpropyl isothiocyanate ≥99%

Supplier: Matrix Scientific

056617-1G 056617-5G
101911-054EA 136.92 USD
101911-054 101911-056
3-Phenylpropyl isothiocyanate ≥99%
3-Phenylpropyl isothiocyanate

Hazard: Irritant-Harmful

Formula: C₁₀H₁₁NS
Boiling Pt: 156…160 °C (12 mmHg)
Density: 1.07
MDL Number: MFCD00041134
CAS Number: 2627-27-2

Order Now

Specification Test Results

Boiling point 156-160°C/1
Density 1.07


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR