Methyl L-2-isothiocyanato-3-methylbutyrate ≥99%

Supplier: Matrix Scientific

056600-1G 056600-5G
101911-004EA 76.92 USD
101911-004 101911-006
Methyl L-2-isothiocyanato-3-methylbutyrate ≥99%
Methyl L-2-isothiocyanato-3-methylbutyrate

Hazard: Irritant-Harmful/Lachrymator

Formula: C₇H₁₁NO₂S
Boiling Pt: 100°C/1mm
Density: 1.07
MDL Number: MFCD09025710
CAS Number: 21055-41-4

Order Now

Specification Test Results

Boiling point 100°C/1mm
Density 1.07

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR