3-Methyl-2-butyl isothiocyanate ≥99%

Supplier: Matrix Scientific

056599-1G 056599-5G
101911-000EA 90.77 USD
101911-000 101911-002
3-Methyl-2-butyl isothiocyanate ≥99%
3-Methyl-2-butyl isothiocyanate

Hazard: Irritant-Harmful

Formula: C₆H₁₁NS
Boiling Pt: 164-167°C
Density: 0.93
MDL Number: MFCD00060388
CAS Number: 201224-92-2

Order Now

Specification Test Results

Boiling point 164-167°C
Density 0.93


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR